20143328 Rev E - Heraeus Megafuge 8 and 8R - User Manual
This centrifuge is a laboratory product used to separate substance mixtures of different densities. This centrifuge can become an In-vitro-diagnostics device (Directive 98/79/EC), if used together with the hematocrit rotor and his accessories. The hematocrit value is determined using the instructions written in the instruction manual of the hematocrit rotor. The instructions are based upon the reference method following DIN 58933, allowing to determinate the erythrocyte volume fraction within
Description:
Use this manual to get acquainted with the centrifuge and its accessories.
Environment:
- Heraeus 8
- Heraeus 8R
Attachment(s):
| File | Last Modified |
|---|---|
| 20143328-e - Heraeus Megafuge 8, 8R Use Instructions.pdf | December 08, 2021 |