P_Program Saving Instructions - AccuSpin 24c
The Fisher Scientific accuSpin 24C Centrifuge is equipped with 3 fixed programs and is able to save 1 additional customizable program. It is only possible to save a program if the centrifuge is not running and the PROGRAMS ONLY mode in the system menu is set to NO.
Description:
This document instructs customers on saving a program to memory.
Environment:
Fisher Scientific accuSpin 24C
Attachment(s):
| File | Last Modified |
|---|---|
| Program Save accuSpin 24C.pdf | December 07, 2021 |