056-720-00 - Thermo Flask Cryogenic Storage - Operating Manual
This manual covers the safe operation of Thermo Fisher Scientific Cryogenics Thermoflasks (models 2122, 2123, 2124, 2129, and 2130). Key points include the importance of wearing PPE when handling liquid nitrogen, ensuring proper ventilation, and pre-cooling flasks to avoid thermal shock. It recommends periodic maintenance and testing every 7-10 years. Specifications and warranty details are also provided.
Description:
Operating, user manual
Environment:
Cryogenics Thermoflasks:
- 2122
- 2123
- 2124
- 2129
- 2130
Attachment(s):
| File | Last Modified |
|---|---|
| 056-720-00 - Rev 1 - Thermo Flask Cryogenic Storage - Operating Manual.pdf | December 06, 2021 |
| 056-720-00 - Thermo Flask Cryogenic Storage - Operating Manual.pdf | December 06, 2021 |