330026H03 - Cardinal Health CHG25FSSA and CHG49FSSA - Installation and Operation manual
The Cardinal Health General Purpose Freezers manual provides installation, operation, safety, and maintenance instructions for models CHG25FSSA and CHG49FSSA. It covers unpacking, setup, wiring, temperature monitoring, alarm settings, control panel use, and maintenance tasks like cleaning and defrosting. It also includes troubleshooting tips, warranty information, and regulatory compliance details.
Description:
Installation and operation manual in English and French.
Environment:
Cardinal Health General Purpose Freezers CHG Series, models:
| Freezers |
| CHG25FSSA |
| CHG49FSSA |
Model Annotation:
| Series | CHG (Cardinal Health) |
| Size (cu.ft) | 25, 49 |
| Type | F = Freezer |
| Exterior | S = Stainless |
| Door Type | S = Solid |
| Voltage | A = 115 V / 60 Hz |
Attachment(s):
| File | Last Modified |
|---|---|
| 330026H03 - Rev B - Cardinal Health CHG25FSSA and CHG49FSSA - Installation and Operation manual.pdf | December 03, 2021 |