7003795 - Forma Laboratory Freezers - Operating and Maintenance Manual
This manual provides operating and maintenance instructions for Thermo Forma Laboratory Freezers, models 3795/3796, 3797/3798, and 3799/3800. It covers installation, startup, operation, and troubleshooting, with safety precautions emphasized. Key features include control panel setup, alarm systems, and minimizing frost build-up. Specifications, parts lists, and warranty information are also included, ensuring proper use and maintenance of the freezers.
Description:
Operating and maintenance manual
Environment:
Thermo Forma Laboratory Freezers, models:
| PART # | FORMA MODEL # | MODEL TYPE | CUBIC FEET | DOORT YPE | VOLTAGE |
| 52-1996-25 | 3795 | Freezer | 27.3 | 1 SolidHinged | 115 |
| 52-1996-26 | 3796 | Freezer | 27.3 | 1 SolidHinged | 220 |
| 52-1996-27 | 3797 | Freezer | 49.1 | 2 SolidHinged | 115 |
| 52-1996-28 | 3798 | Freezer | 49.1 | 2 SolidHinged | 220 |
| 52-1996-29 | 3799 | Freezer | 76.2 | 3 SolidHinged | 115 |
| 52-1996-30 | 3800 | Freezer | 76.2 | 3 SolidHinged | 220 |
| 52-1984-31 | 3801 | Cold Wall Freezer | 27.3 | 1 SolidHinged | 115 |
| 52-1984-32 | 3802 | Cold Wall Freezer | 27.3 | 1 SolidHinged | 220 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7003795 - Rev 3 - Forma Laboratory Freezers - Operating and Maintenance Manual.pdf | December 03, 2021 |