7018097 - ThermoForma 8097 and 8103 Plasma Freezers - Operating and Maintenance Manual
This manual provides operating and maintenance instructions for Thermo Forma Plasma Freezers, models 8097 and 8103. It includes guidance on installation, temperature setting, alarm connections, diagrams, parts list, and maintenance. Key features include built-in temperature alarms and 7-day chart recorders. Regular maintenance ensures optimal performance. For assistance, contact Thermo Forma support.
Description:
Operating and maintenance manual. Models 8097 (120V, 60Hz) and 8103 (220V, 50Hz) are Thermo Forma Plasma Freezers with built-in, over/under temperature alarm systems and 7-day chart recorders as required by regulatory agencies for plasma storage. The freezer operates at - 30°C (-22°F).
Environment:
ThermoForma 19.7 cu. ft. capacity Plasma Freezer:
- Model 8097 120V, 60 Hz
- Model 8103 220V, 50Hz
Attachment(s):
| File | Last Modified |
|---|---|
| 7018097 - Rev 4 - ThermoForma 19.7 cu. ft. Capacity Plasma Freezer - Operating and Maintenance Manual.pdf | December 10, 2021 |