7048800 - Thermo Scientific Forma 7000 Series -40C Upright Laboratory Freezer - Operating and Maintenance Manual
The Thermo Scientific Forma 7000 Series -40C Upright Laboratory Freezer User Manual covers catalog numbers 7240A, 7240V, 7240D, 7320A, 7320V, 7320D, 7400A, 7400V, and 7400D. It provides installation, operation, maintenance instructions, schematics, drawings, and exploded parts view with part numbers. Key sections include safety guidelines, calibration of the control probe, alarm settings, and maintenance tasks like cleaning filters and condensers.
Description:
Operating and maintenance manual.
Environment:
Thermo Scientific Forma 7000 Series -40C Upright Laboratory Freezer catalog numbers and models:
| Catalog Number | Model | Capacity in Cubic Feet | Voltage |
| 7240A | 8800 | 13 | 120/60 |
| 7240V | 8801 | 13 | 230/50 |
| 7240D | 8803 | 13 | 230/60 |
| 7320A | 8804 | 17 | 120/60 |
| 7320V | 8806 | 17 | 230/50 |
| 7320D | 8807 | 17 | 230/60 |
| 7400A | 8809 | 23 | 120/60 |
| 7400V | 8810 | 23 | 230/50 |
| 7400D | 8811 | 23 | 230/60 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7048800 - Rev 1 - Thermo Scientific Forma 7000 Series -40C Upright Laboratory Freezer - Operating and Maintenance Manual.pdf | December 03, 2021 |