102999 - Rev A - Fisher Scientific Isotemp Laboratory Freezers - User Manual
This article is a detailed guide to Fisher Scientific's Isotemp Laboratory Freezers, covering models 13-986-223F/FA/FR, 13-986-247F/FA/FR, and 13-986-274F/FA/FR. It includes safety information, unpacking instructions, performance characteristics, installation guidelines, operational procedures, troubleshooting tips, maintenance requirements, replacement parts, wiring diagrams, and warranty details, emphasizing the importance of proper understanding for safe operation.
Description:
User manual
Environment:
| Catalog Number | Volts(±10%) | Amps | Hz |
| 13-986-223F 13-986-223FA 13-986-223FR |
115 | 10 | 60 |
| 13-986-247F 13-986-247FA 13-986-247FR |
230 | 10.5 | 60 |
| 13-986-274F 13-986-274FA 13-986-274FR |
230 | 16 | 60 |
| 13-986-223FHK 13-986-223FRHK 13-986-223FAHK |
220 | 6.2 | 50 |
| 13-986-247FHK 13-986-247FRHK 13-986-247FAHK |
220 | 6.2 | 50 |
| 13-986-274FHK 13-986-274FRHK 13-986-274FAHK |
220 | 6.2 | 50 |
Attachment(s):
| File | Last Modified |
|---|---|
| 102999 - Rev A - Fisher Scientific Isotemp Laboratory Freezers - User manual.pdf | December 03, 2021 |