326806H01 - Fisher Scientific Explosion Proof Refrigerators and Freezers - User manual
This guide covers Fisher Scientific's Isotemp Explosion-Proof Refrigerators (model 13-986-153) and Freezers (model 13-986-150). It includes safety information, unpacking instructions, installation guidelines, operation procedures, maintenance tips, troubleshooting advice, and warranty details. Emphasizing proper use for safe operation in hazardous locations, it details electrical requirements, leveling, temperature control, and sealing the Killark box to prevent explosions.
Description:
User manual
Environment:
Isotemp® Explosion-Proof Refrigerators and Freezers, models:
- Exp. Refrigerator : 13-986-153 (20EREEFSA)
- Exp. Freezer : 13-986-150 (20EFEEFSA)
| Model | Total Vol. Cu. Ft. | Exterior Dimensions H x W x D inches | Electrical Characteristics Volts/Hz, Amps | Temp. Range °C(° F) |
| 13-986-153 | 21 | 73.25" X 34" X 35.81" (186.1 X 86.4X91.0 cm) | 115/60, 5.0 | 1" to 12°(34° to 54°) |
| 13-986-150 | 21 | 73.25" X 34" X 35.81" (186.1 X 86.4X91.0 cm) | 115/60, 5.0 | 12° to -20°(-4° to 10°) |
Attachment(s):
| File | Last Modified |
|---|---|
| 326806H01 - Rev D - Fisher Scientific Explosion Proof Refrigerators and Freezers - User manual.pdf | December 03, 2021 |