326629H01 - Rev B - Thermo Scientific Jewett Pass-Thru Blood Bank Refrigerators - Installation and Operation Manual
This manual provides installation and operation instructions for Thermo Scientific Jewett Pass-Thru Blood Bank Refrigerators. It covers specifications, safety precautions, unpacking, installation, wiring, operation, drawer management, hinge and latch adjustments, maintenance, troubleshooting, and accessories. The guide emphasizes proper procedures for optimal performance, safety, and longevity.
Description:
Installation and operation manual
Environment:
Thermo Scientific Jewett® Pass-Thru Blood Bank Refrigerators, models:
| Model | Voltage | Frequency | Current |
| BBR55D-1B | 120 V | 60 HZ | 12 A |
| BBR55D-2A* | 230 V | 50 HZ | 6.1 A |
| BBR25D-1B | 120 V | 60 HZ | 9 A |
| BBR25D-2A* | 230 V | 50 HZ | 4.5 A |
* Not CE marked
Attachment(s):
| File | Last Modified |
|---|---|
| 326629H01 - Rev B - Thermo Scientific Jewett Pass-Thru Blood Bank Refrigerators - Installation and Operation Manual.pdf | December 03, 2021 |