7028813 - Thermo Scientific Revco DxF -40C Upright Laboratory Freezer - Operating Manual
Thermo Scientific Revco DxF -40°C Upright Laboratory Freezer manual provides installation, operation, calibration, alarm, and preventive maintenance guidance. Covers temperature and alarm setpoints, remote alarm contacts, control probe calibration, error codes, air filter and condenser cleaning, defrosting, vacuum relief port care, batteries, optional CO2/LN2 backup system, chart recorder, datalogger, and freezer specifications.
Description:
Operating manual
Environment:
Thermo Scientific Revco® DxF -40C Upright Laboratory Freezer, catalog numbers and models:
| Catalog Number | Model | Capacity - Cubic Feet | Voltage |
| DxF24040A | 8813 | 13 | 120/60 |
| DxF24040V | 8814 | 13 | 230/50 |
| DxF24040D | 8816 | 13 | 230/60 |
| DxF32040A | 8817 | 17 | 120/60 |
| DxF32040V | 8819 | 17 | 230/50 |
| DxF32040D | 8820 | 17 | 230/60 |
| DxF40040A | 8821 | 23 | 120/60 |
| DxF40040V | 8822 | 23 | 230/50 |
| DxF40040D | 8823 | 23 | 230/60 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7028813 - Rev 0 - Thermo Scientific Revco DxF -40C Upright Laboratory Freezer - Operating Manual..pdf | September 15, 2026 |
| 7028813 - Rev 1 - Thermo Scientific Revco DxF -40C Upright Laboratory Freezer - Operating Manual.pdf | December 03, 2021 |