325099H11 - -30 Laboratory and Plasma Freezers A and D Voltages - Installation and Operation Manual
Description:
This manual provides installation and operation instructions for laboratory and plasma freezers with a preset temperature setpoint of -30°C.
Environment:
-30°C Laboratory and Plasma Freezers A and D, models:
Model number | Voltage/Frequency | Amperage | Plug Type | Power Module Plug |
430A 430D |
115 208/230 |
10 7 |
5-15 P 6-15 P |
IEEC19 |
1230A 1230D |
115 208/230 |
12 | 5-15 P 6-15 P |
IEEC19 |
2330A 2330D |
115 208/230 |
12 8 |
5-15 P 6-15 P |
IEEC19 |
3030A 3030D |
115 208/230 |
15.5 12 |
5-20 P 6-15 P |
IEEC19 |
5030A 5030D |
115 208/230 |
16 12 |
5-20 P 6-15 P |
IEEC19 |
Model numbers:
FFGL430 | FFGL1230 | FFGL2330 | IUF3004 | FFGL3030 | IUF3050 | FFGL5030 |
FFPF430 | FFPF1230 | FFPF2330 | IUF3012 | FFPF3030 | FFPF5030 | |
JLF430 | JLF1230 | FFPH2330 | IUF3023 | IUF3030 | JLF5030 | |
JPL430 | PPL1230 | JLF2330 | JLF3030 | JPL5030 | ||
PPL430 | UFP1230 | JPL2330 | JPL3030 | PPL5030 | ||
UFP430 | ULT1230 | PPL2330 | UFP3030 | UFP5030 | ||
ULT430 | UFP2330 | ULT3030 | ULT5030 | |||
ULT2330 |