Thermo Fisher Scientific
332192H01 - Rev A - -20C Flammable Materials Storage Laboratory Freezers TSX Series Installation and Operation Manual
Description: 
This manual provides installation and operation instructions for Flammable Material Storage (FMS) laboratory freezers with a preset temperature setpoint of -20°C.
Environment:
-20° C Flammable Materials Storage Laboratory Freezers TSX Series, model TSX2320HA
Unit Specifications:
Model Rated Voltage Rated Current Frequency / Phase Power Module Plug Approximate Lab Freezer Weight Kg (lbs) Approximate Freezer Weight Kg (lbs) Exterior Dimensions (D x W x H)   Hydrocarbon
23A 115 V 5.56 A 60 Hz IEC C19 178 (394) 203 (449) 93.6 x 71.1 x 199.4 cm (36.9 x 28.0 x 78.5 in) R290, 145 g