333772H01 - Isotemp Freezer - Installation and Operation Manual
This article provides installation and operation instructions for the Thermo Scientific General Purpose Freezer model TSV18FPSA. It details safety information, product specifications, unpacking guidelines, operating conditions, and installation steps. It also includes operation instructions, maintenance tips, troubleshooting, and warranty information. The freezer is designed for laboratory use, storing samples between -22°C and -10°C, and is not suitable for medical diagnostic samples.
Description:
The following is installation operation manual (user manual) for model TSV18FPSA.
Environment:
TSV18FPSA, TSV18FPSA user manual, TSV18FPSA guide
Attachment(s):
| File | Last Modified |
|---|---|
| 333772H01 - Isotemp Freezer - Installation and Operation Manual Rev B.pdf | November 21, 2025 |