329688H01 - Rev B - VWR General Purpose Refrigerators and Freezer - Installation and Operation Manual
Description:
These are general-purpose units available in all refrigerator, freezer and combination refrigerator/freezer models.
Environment:
VWR General Purpose Refrigerators and Freezer, models:
| 17LREEVWA | 18LCEEVWA | 25LCEEVWA | |
| Size (Cu. Ft.) | 17 | 18 | 25 |
| Shipping Weight | 190 lbs (86.2 kg) | 205 lbs (93 kg) | 315 1b (142.8 kg) |
| Plug Type | NEMA 5-15P | NEMA 5-15P | NEMA 5-15P |
| Electric Power | 115V/60 Hz | 115 V/60 Hz | 115 V/60 Hz |
| Rated Current (Amps) | 5 | 6 | 11.6 |
| Defrost | Manual | Automatic | Automatic |
Attachment(s):
| File | Last Modified |
|---|---|
| 329688H01 - Rev B - VWR General Purpose Refrigerators and Freezer - Installation and Operation Manual.pdf | December 14, 2021 |