34830H56 - Rev B - Cardinal Health Gold Series Blood Bank Refrigerators - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Cardinal Health Gold Series Blood Bank Refrigerators Installation and Operation Manual. This manual provides installation and operation instructions for: Undercounter models, Space Saver models and Upright models:
Voltage Codea | Amps/Breaker | Cu. Ft./Liters | Cabinet |
A | 6.0/15 | 4.7/133 | Undercounter |
D | 6.0/15 | 4.7/133 | Undercounter |
V, W | 6.0/15 | 4.7/133 | Undercounter |
A | 9.4/15 | 11.5/326 | Space Saver |
D | 6.0/15 | 11.5/326 | Space Saver |
V, W | 6.0/15 | 11.5/326 | Space Saver |
A | 9.4/15 | 23.3/659 | Upright |
D | 6.0/15 | 23.3/659 | Upright |
V, W | 6.0/15 | 23.3/659 | Upright |
A | 9.4/15 | 29.2/826 | Upright |
D | 6.0/15 | 29.2/826 | Upright |
V, W | 6.0/15 | 29.2/826 | Upright |
A | 15.0/20 | 51.1/1447 | Upright |
D | 8.5/15 | 51.1/1447 | Upright |
V, W | 8.5/15 | 51.1/1447 | Upright |
a V = 230V/50Hz CEE7/7 Plug Cap (Schuko)
W = 230V/50Hz BS1363a Plug Cap
Attachment(s):
File | Last Modified |
---|---|
34830H56 - Rev B - Cardinal Health Gold Series Blood Bank Refrigerators - Installation and Operation Manual.pdf | December 10, 2021 |