34830H52 - Rev B - Thermo Scientific Puffer Hubbard Blood Bank Refrigerators - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Thermo Scientific Puffer Hubbard Blood Bank Refrigerators. This manual provides installation and operation instructions for undercounter models, space saver models and upright models:
| Model | Voltage Codea | Amps/ Breaker | Cu. Ft./Liters | Cabinet |
| HLT-4V-4BB | A | 6.0/1.5 | 4.7/133 | Undercounter |
| HLT-4V-4BB | D | 6.0/15 | 4.7/133 | Undercounter |
| HLT-4V-4BB | V, W | 6.0/15 | 4.7/133 | Undercounter |
| HLT-12V-4BB | A | 9.4/15 | 11.5/326 | Space Saver |
| HLT-12V-4BB | D | 6.0/15 | 11.5/326 | Space Saver |
| HLT-12V-4BB | V, W | 6.0/15 | 11.5/326 | Space Saver |
| HLT-23V-4BB | A | 9.4/15 | 23.3/659 | Upright |
| HLT-23V-4BB | D | 6.0/15 | 23.3/659 | Upright |
| HLT-23V-4BB | V, W | 6.0/15 | 23.3/659 | Upright |
| HLT-30V-4BB | A | 9.4/15 | 29.2/826 | Upright |
| HLT-30V-4BB | D | 6.0/15 | 29.2/826 | Upright |
| HLT-30V-4BB | V, W | 6.0/15 | 29.2/826 | Upright |
| HLT-50V-4BB | A | 15.0/20 | 51.1/1447 | Upright |
| HLT-50V-4BB | D | 8.5/15 | 51.1/1447 | Upright |
| HLT-50V-4BB | V, W | 8.5/15 | 51.1/1447 | Upright |
a V = 230V/50Hz CEE7/7 Plug Cap (Schuko)
W = 230V/50Hz BS1363a Plug Cap
Attachment(s):
| File | Last Modified |
|---|---|
| 34830H52 - Rev B - Thermo Scientific Puffer Hubbard Blood Bank Refrigerators - Installation and Operation Manual.pdf | December 10, 2021 |