34830H21 - Rev J - Revco Blood Bank Refrigerators - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
This manual provides installation and operation instructions for undercounter models, space saver models and upright models:
| Model | Voltage Codea | Amps/ Breaker | Cu. Ft./ Liters |
Cabinet |
| REB-404 | A | 6.0/15 | 4.7/ | Undercounter |
| REB-404 | D | 6.0/15 | 4.7/ | Undercounter |
| REB-404 | V, W | 6.0/15 | 4.7/ | Undercounter |
| REB-1204 | A | 9.4/15 | 11.5/326 | Space Saver |
| REB-1204 | D | 6.0/15 | 11.5/326 | Space Saver |
| REB-1204 | V, W | 6.0/15 | 11.5/326 | Space Saver |
| REB-2304 | A | 9.4/15 | 23.3/659 | Upright |
| REB-2304 | D | 6.0/15 | 23.3/659 | Upright |
| REB-2304 | V, W | 6.0/15 | 23.3/659 | Upright |
| REB-3004 | A | 9.4/15 | 29.2/826 | Upright |
| REB-3004 | D | 6.0/15 | 29.2/826 | Upright |
| REB-3004 | V, W | 6.0/15 | 29.2/826 | Upright |
| REB-5004 | A | 15.0/20 | 51.1/1447 | Upright |
| REB-5004 | D | 8.5/15 | 51.1/1447 | Upright |
| REB-5004 | V, W | 8.5/15 | 51.1/1447 | Upright |
a A = 115V, 60Hz; D = 208/230V, 60Hz; V = 220V, 50Hz; W = 240V, 50 Hz
Attachment(s):
| File | Last Modified |
|---|---|
| 34830H21 - Rev J - Revco Blood Bank Refrigerators - Installation and Operation Manual.pdf | December 10, 2021 |