324019H01 - Thermofisher Scientific MR05 Series Undercounter Lab Refrigerator - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Thermo Scientific MR05 Series Undercounter Laboratory Refrigerators:
- MR05PA
- MR05PS-SEEE
- MR05PA-SEEE-TS
| Model Series | MR05 |
| Model Style | Undercounter |
| Capacity | 5.4 cu ft (153 Liters) |
| Exterior Dimensions: | |
| Height (inches) | 34 5/8" |
| Width (inches) | 24" |
| Depth (inches) | 24 1/8" Cabinet Depth |