328277H0x - Thermo Scientific TSX505 - Pass Code Addendum
Description:
Addendum for the section 8.6.1 of the manual. During setup, use the code 3265 to open the door for first time. We recommend that you change the log in code during initial set up for enhanced security.
Environment:
- TSX505G
- TSX505S
- TSX505GA
- TSX505GD
- TSX505GV
- TSX505GW
- TSX505GC
- TSX505SA
- TSX505SD
- TSX505SV
- TSX505SW
- TSX505SAC
Attachment(s):
| File | Last Modified |
|---|---|
| 328277H0x - Thermo Scientific TSX505 - Pass Code Addendum.pdf | December 02, 2021 |