7000902 - Forma 900 Series ULT Freezers - User Manual
Description:
Operating and maintenance manual, revisions 0 and 1.
Environment:
Thermo Forma 900 Series Ultra Low Temperature Upright Freezers
Models | Capacity in Cubic Feet | Voltage |
902 | 13 | 230 |
903 | 13 | 120 |
904 | 17 | 120 |
905 | 17 | 230 |
906 | 23 | 230 |
907 | 28 | 230 |
Double Door Units | ||
991 | 13 | 230 |
992 | 13 | 120 |
993 | 17 | 120 |
994 | 17 | 230 |
995 | 23 | 230 |
Attachment(s):
File | Last Modified |
---|---|
7000902 - Rev 1 - Forma 900 Series ULT Freezers - User Manual.pdf | November 22, 2021 |
7000902 - Rev 0 - Forma 900 Series ULT Freezers - User Manual.pdf | November 22, 2021 |