7038602 - Forma 8600 Series ULT Freezer - User Manual
Description:
Operating and maintenance manual
Environment:
Model 8600 Series -86C Ultra Low Temperature Freezer
| Model | Capacity in Cubic Feet | Voltage |
| 8602 | 13 | 230 |
| 8603 | 13 | 120 |
| 8604 | 17 | 120 |
| 8605 | 17 | 230 |
| 8606 | 23 | 230 |
| 8607 | 28 | 230 |
| 8656 | 23 | 120 |
| Double Door Units | ||
| 8690 | 23 | 120 |
| 8691 | 13 | 230 |
| 8692 | 13 | 120 |
| 8693 | 17 | 120 |
| 8694 | 17 | 230 |
| 8695 | 23 | 230 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7038602 - Rev 0 - Forma 8600 Series ULT Freezer - User Manual.pdf | November 24, 2021 |
| 7038602 - Rev 3 - Forma 8600 Series ULT Freezer - User Manual.pdf | November 24, 2021 |
| 7038602 - Rev 4 - Forma 8600 Series ULT Freezer - User Manual.pdf | November 24, 2021 |
| 7038602 - Rev 5 - Forma 8600 Series ULT Freezer - User Manual.pdf | November 24, 2021 |
| 7038602 - Rev 1 - Forma 8600 Series ULT Freezer - User Manual.pdf | November 24, 2021 |
| 7038602 - Rev 2 - Forma 8600 Series ULT Freezer - User Manual.pdf | November 24, 2021 |
| 7038602 - Rev 6 - Forma 8600 Series ULT Freezer - User Manual.pdf | November 22, 2021 |