Revco Ultra Low Temperature Freezers - Check Fuse Warning Instruction Sheet
Description:
Instruction sheet for checking the surge suppressor. The message "check fuse" typically indicates the fuse in the freezer’s built in surge protector has failed.
Environment:
Revco ultra low temperature freezers with built in surge suppressor
ULT350-9-A | ULT1090-9-A | ULT1490-9-A | ULT1790-9-A | ULT2186-9-A | ULT3286-9SI-D |
ULT350-9-D | ULT1090-9-D | ULT1490-9-D | ULT1790-9-D | ULT2186-9-D | ULT3286-9SI-V |
ULT350-9-V | ULT1090-9-V | ULT1490-9-V | ULT1790-9-V | ULT2186-9-V | ULT3286-9SI-W |
ULT350-9-W | ULT1090-9-W | ULT1490-9-W | ULT1790-9-W | ULT2186-9-W | |
ULT390-9-A | ULT1340-9-A | ULT1740-9-A | ULT2050-9-A | ULT2540-9-A | |
ULT390-9-D | ULT1340-9-D | ULT1740-9-D | ULT2050-9-D | ULT2540-9-D | |
ULT390-9-V | ULT1340-9-V | ULT1740-9-V | ULT2050-9-V | ULT2540-9-V | |
ULT390-9-W | ULT1340-9-W | ULT1740-9-W | ULT2050-9-W | ULT2540-9-W | |
ULT750-9-A | ULT1386-9-A | ULT1750-9-A | ULT2090-9-A | ULT2586-9-A | |
ULT750-9-D | ULT1386-9-D | ULT1750-9-D | ULT2090-9-D | ULT2586-9-D | |
ULT750-9-V | ULT1386-9-V | ULT1750-9-V | ULT2090-9-V | ULT2586-9SI-A | |
ULT750-9-W | ULT1386-9-W | ULT1750-9-W | ULT2090-9-W | ULT2586-9SI-D | |
ULT1050-9-A | ULT1450-9-A | ULT1786-9-A | ULT2140-9-A | ULT2586-9SI-V | |
ULT1050-9-D | ULT1450-9-D | ULT1786-9-D | ULT2140-9-D | ULT2586-9SI-W | |
ULT1050-9-V | ULT1450-9-V | ULT1786-9-V | ULT2140-9-V | ULT2586-9-V | |
ULT1050-9-W | ULT1450-9-W | ULT1786-9-W | ULT2140-9-W | ULT2586-9-W |
Attachment(s):
File | Last Modified |
---|---|
Revco Ultra Low Temperature Freezers - Check Fuse Warning Instruction Sheet.pdf | November 22, 2021 |