334555H01 - TSX Core ULT Freezers - User Manual
The Ultra Low Temperature Freezers TSX Core are designed for long-term storage at -50°C to -86°C. This manual covers installation, operation, and maintenance. Key features include EPA ENERGY STAR compliance, advanced door operation, and safety precautions. Regular maintenance involves cleaning the condenser, defrosting the freezer, and checking the battery. The troubleshooting guide addresses common issues. Warranty details and end-of-life care instructions are provided.
Description:
Installation and Operation Manual also known as user manual for TSX Core ULT Freezer Series.
Environment:
TSX40086CA , TSX50086CA,TSX60086CA, TSX70086CA
Attachment(s):
| File | Last Modified |
|---|---|
| 334555H01 - TSX Core ULT Freezers - User Manual Rev. B Dec 2024.pdf | April 17, 2025 |