7055608 - VWR - ULT Chest Freezer Instruction Manual
Description:
VWR® ULT Chest Freezers Instruction Manual
Environment:
VWR® ULT Chest Freezers Instruction Manual catalogue and model numbers:
North American Catalogue Number | European Catalogue Number | Model Number | Size (cu ft) | Size (liters) | Box | Voltage |
-86°C FREEZERS | ||||||
10160-738 | -- | VWRC0386A | 3 | 85 | 63 | 120V/60Hz |
-- | 471-1137 | VWRC0386V | 3 | 85 | 63 | 230/50Hz |
10160-740 | VWRC1386A | 13 | 368 | 252 | 120V/60Hz | |
10160-742 | -- | VWRC1386D | 13 | 368 | 252 | 208-230V/60Hz |
-- | 471-1138 | VWRC1386V | 13 | 368 | 252 | 230/50Hz |
10160-744 | -- | VWRC2086A | 20 | 566 | 396 | 120V/60Hz |
10160-746 | -- | VWRC2086D | 20 | 566 | 396 | 208-230V/60Hz |
-- | 471-1139 | VWRC2086V | 20 | 566 | 396 | 230/50Hz |
Attachment(s):
File | Last Modified |
---|---|
7055608 - Rev 7 - VWR - ULT Chest Freezer Manual.pdf | November 22, 2021 |
7055608 - Rev 10 - VWR - ULT Chest Freezer Manual.pdf | November 19, 2021 |