Thermo Scientific - Testing of Self Powered Temperature Transmitters Red Minco RoHS Instruction Sheet
Description:
Testing of Self Powered 4-20mA Temperature Transmitter Options
Option #s 7203-AV, 7203FMS, 7204, 7205TA (or 4-20mA loops in general)
Environment:
7203-AV, 7203FMS, 7204, 7205TA (or 4-20mA loops in general)
- Refrigerators
- Freezers
- ULT Freezers
- Main parts– 326518H01 through H04 TempTrans (Minco TT518)
- 327624H01 24v DC Power Supply or 16.5v DC PowerSupply and a dummyload resistor ( 1/4w 5%)– 25 ohms or 250 ohms Refer to instructional drawing #s 311977I02
- 325436H01 FMS and wiring diagram #s 325439H01 FMS
- General 326861H01
- 327987H01 TSX.
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific - Testing of Self Powered Temperature Transmitters Red Minco RoHS Instruction Sheet.pdf | November 22, 2021 |