Thermo Fisher Scientific
Thermo Scientific 7400 CryoPlus Series - How to Reset Access Code Instructions
This article explains how to reset access codes on Thermo Fisher Scientific CryoPlus Series 7400 models. It covers locking the key switch, generating a 6-digit seed, calculating a 4-digit service password, entering it to access the EE menu, confirming the reset, restoring normal operation, and setting/verifying the customer passcode. Applies to CryoPlus 1–4 Medical Device and General Purpose units.
Description: 
When customers loses Access Code a factory reset would have to be performed. The attached provides full step by step Instructionsopn completing this process. 
Environment:
Model
Series
7400
CryoPlus 1 Medical Device
7401
CryoPlus 1 Medical Device
7401GP
CryoPlus 1 General Purpose
7402
CryoPlus 2 Medical Device
7403
CryoPlus 2 Medical Device
7403 GP
CryoPlus 2 General Purpose
7404
CryoPlus 3 Medical Device
7405
CryoPlus 3 Medical Device
7405 GP
CryoPlus 3 General Purpose
7406
CryoPlus 4 Medical Device
7407
CryoPlus 4 Medical Device
7407 GP
CryoPlus 4 General Purpose
  • Was this article helpful?