Thermo Scientific CryoMed Controlled Rate Freezer with OPC UA Communication
This article provides a detailed guide on installing and configuring the UaExpert software to establish OPC UA communication with Thermo Scientific™ CryoMed™ Controlled-Rate Freezer. It covers the necessary prerequisites, software installation steps, and procedures for setting up and verifying two-way communication between UaExpert and CRF. The instructions include downloading and installing UaExpert, accessing CRF settings, connecting via Endpoint URL, and confirming data reading and writing co
Description:
The intent of the Control Rate Freezer OPC UA is to provide a means for the customer to communicate, monitor and have limited control of the Control Rate Freezers. This instruction will illustrate how to test the communication.
Environment:
- TSCM17SA
- TSCM17FA
- TSCM17SV
- TSCM17FV
- TSCM34SA
- TSCM34FA
- TSCM34SV
- TSCM34FV
- TSCM48SA
- TSCM48FA
- TSCM48SV
- TSCM48FV
- TSCM17CA
- TSCM17TA
- TSCM34CA
- TSCM34TA
- TSCM48CA
- TSCM48TA
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific CryoMed Controlled-Rate Freezer w OPC UA Revision 1.pdf | September 21, 2022 |
| Thermo Scientific™ CryoMed™ Controlled-Rate Freezer w OPC UA.pdf | March 25, 2022 |