+4 -20 -30C IntrLogic Refrigerator and Freezer - Resetting Factory Default Programming and Service Settings Instruction Sheet
The article provides instructions for resetting factory default programming and service settings on IntrLogic Refrigerators/Freezers at temperatures of +4°, -20°, and -30°C. Reasons for resetting include improper unit control behavior, troubleshooting, saving time, and new board installation. The reset process involves turning the key switch off, holding the up and down arrows while turning the switch on, and verifying default settings. Additionally, it lists specific models applicable to these
Description:
How to reset factory default programming and service settings
Environment:
+4°, -20° and -30°C High Performance Refrigerator and Freezer.
| UEN2320 | RFMS2320 | |
| UEN3020 | PPL1230 | JPL1230 |
| UFP1230 | PPL2330 | JPL2330 |
| UFP2330 | PPL3030 | JPL3030 |
| UFP3030 | LF2330 | JPL430 |
| UFP430 | LF3030 | JPL5030 |
| UFP5030 | LF430 | IEN2023 |
| UGL2320 | LF5030 | IEN2030 |
| UGL3020 | IUF3012 | IGL2023 |
| ULT1230 | IUF3023 | IGL2030 |
| ULT2330 | IUF3030 | FFMS2320 |
| ULT3030 | IUF3050 | FFPF1230 |
| ULT430 | JLF1230 | FFPF2330 |
| ULT5030 | JLF2330 | FFPF3030 |
| UFP1230 | JLF3030 | FFPF430 |
| UPF2330 | JLF430 | FFPF5030 |
| UPF3030 | JLF5030 | FFGL430 |
| UPF430 | LF1230 | FFGL2320 |
| UPF5030 | FFGL5030 | FFGL2330 |
| FFEN2320 | FFEN3020 | FFGL3020 |
| TSFMS2320 | FFGL1230 | FFGL3030 |
| PPL5030 |
Attachment(s):
| File | Last Modified |
|---|---|
| +4 -20 -30C IntrLogic Refrigerator_Freezer - Resetting Factory Default Programming and Service Settings Instruction Sheet.pdf | December 06, 2021 |