TSG CHG FBG General Purpose Freezer - How to adjust temperature setpoint on Carel Controller Instructions
This article explains how to adjust the temperature setpoint on Carel controllers for TSG, CHG, and FBG freezers. Steps include accessing the controller menu, entering the parameter menu, locating the setpoint parameter, changing the setpoint, and saving the new settings. Key actions involve pressing the Program button, using arrow keys, and entering a default password if needed. Wait 5 seconds after changes before powering off to save settings.
Description:
The attachment provides procedure on adjusting temperature set point, also known as control set point on Carel Controller.
Environment:
- TSG1225FA
- TSG2325FA
- TSG5025FA
- FBG1225FA
- FBG2325FA
- FBG5025FA
- CHG1225FA
- CHG2325FA
- CHG5025FA
Attachment(s):
| File | Last Modified |
|---|---|
| TSG CHG FBG General Purpose Freezer -How to adjust temp setpoint on Carel Control Instructions.pdf | November 10, 2025 |