TSU FBU CHU 405 Series – How to Change Temperature from Celsius to Fahrenheit - Instruction Sheet
This article explains how to switch the temperature display from Celsius to Fahrenheit on the TSU405 Refrigerator Series via the Service Menu. It covers entering password 255, navigating to Parameters (s5) and P07, changing flag 106 to 107, and returning to the main screen. Keywords: TSU405, temperature units, Fahrenheit, Celsius, Service Menu, P07 parameter, appliance settings, user interface
Description:
The attached instructions or procedure provide step by step guidance on changing the TSU405SA or TSU405GA Temperature readout from Celsius to Farenheit.
Environment:
TSU405, FBU405, CHU405, undercounter refrigerator
| TSU405GA | FBU405GA | CHU405GA |
| TSU405SA | FBU405SA | CHU405SA |
Attachment(s):
| File | Last Modified |
|---|---|
| TSU FBU CHU 405 Series – How to Change Temperature from Celsius to Fahrenheit.pdf | January 22, 2026 |