TSG205SA Countertop Refrigerator Series - How to Replace Battery
To replace the battery on the TSG205SA, follow these steps: 1. Locate the battery compartment on the lower rear of the left side panel. 2. Remove the two screws securing the pass-through port and access panel. 3. Remove the panel. 4. Remove and replace only with a CR123A or IEC16340 type battery. 5. Replace the panel and tighten the two screws. We recommend that you keep spare batteries on hand to continue to monitor internal temperature in the event of a power outage.
Description:
Attached are instructions or directions on changing battery when battery is low.
Environment:
TSG205SA, 1.8 cu.ft. Countertop Refrigerator
Attachment(s):
| File | Last Modified |
|---|---|
| TSG205SA Series - How to Replace Battery .pdf | December 27, 2023 |