TSX505 Undercounter Refrigerator Series Instructions – How to Replace the Battery
This article provides step-by-step instructions for replacing the battery in the TSX505 Undercounter Refrigerator Series. It details the process of opening the I/O door, removing and replacing the battery with a CR123A or IEC16340 type, and closing the I/O door. This guide is essential for maintaining the refrigerator's functionality and ensuring proper operation.
Description:
Attachment provides instructions on replacing the TSX505 and CAX undercouter refrigerator Series.
Environment:
- Product: TSX505 Undercounter Refrigerator Series
- TSX505
- TSX505SA
- TSX505GA
- TSX505SD
- TSX505GD
- TSX505SV
- TSX505GV
- TSX505SC
- TSX505GC
- TSX505SW
- TSX505GW
- CA505SA
- CA505GA
- CA505SD
- CA505GD
- battery installation
Attachment(s):
| File | Last Modified |
|---|---|
| Instructions - TSX505 Undercounter Refrigerator Series – How to Replace the Battery.pdf | July 24, 2025 |