PEEK Series ULT Freezers - How to Check Event Log Instruction Sheet
This guide explains how to check the event log on PEEK Series ULT Freezers. First, press the blue, yellow, or red heart icon on the home screen to access the Health System Status Overview. Next, select the event notepad with a pen icon to view a detailed list of events by date and time. The event log table will appear, allowing you to view all items or use icon filters to narrow down specific events, such as door opening activity.
Description:
How to check event log on PEEK Series ULT Freezers.
Environment:
PEEK Series event table.
| UxF30086A | HFU300TA | 88300A | TSU300A |
| UxF30086D | HFU300TD | 88300D | TSU300D |
| UxF30086V | HFU300TV | 88300V | TSU300V |
| UxF40086A | HFU400TA | 88400A | TSU400A |
| UxF40086D | HFU400TD | 88400D | TSU400D |
| UxF40086V | HFU400TV | 88400V | TSU400V |
| UxF50086A | HFU500TA | 88500A | TSU500A |
| UxF50086D | HFU500TD | 88500D | TSU500D |
| UxF50086V | HFU500TV | 88500V | TSU500V |
| UxF60086A | HFU600TA | 88600A | TSU600A |
| UxF60086D | HFU600TD | 88600D | TSU600D |
| UxF60086V | HFU600TV | 88600V | TSU600V |
| UxF70086D | HFU700TD | 88700D | TSU700D |
| UxF70086V | HFU700TV | 88700V | TSU700V |
Attachment(s):
| File | Last Modified |
|---|---|
| Instructions - PEEK Series ULT Freezers - How to Check Event Log.pdf | June 09, 2023 |