325249I01 - Thermo Scientific PEEK - CO2 Back Up System Installation Instructions
Description:
PEEK ULT Freezer CO2 BUS field installation instructions for catalog number FCO4567
Environment:
Revco UXF, Forma 88000, Herasafe HFU and Thermo TSU Series Freezers CO2 backup system.
These instructions are for all PEEK freezers of model size 400 to 700. The 300 size freezer accepts a freestanding BUS only.
| UxF40086A | HFU400TA | 88400A | TSU400A |
| UxF40086D | HFU400TD | 88400D | TSU400D |
| UxF40086V | HFU400TV | 88400V | TSU400V |
| UxF50086A | HFU500TA | 88500A | TSU500A |
| UxF50086D | HFU500TD | 88500D | TSU500D |
| UxF50086V | HFU500TV | 88500V | TSU500V |
| UxF60086A | HFU600TA | 88600A | TSU600A |
| UxF60086D | HFU600TD | 88600D | TSU600D |
| UxF60086V | HFU600TV | 88600V | TSU600V |
| UxF70086D | HFU700TD | 88700D | TSU700D |
| UxF70086V | HFU700TV | 88700V | TSU700V |
Attachment(s):
| File | Last Modified |
|---|---|
| 325249I01_RevE_PEEK_CO2 Nov 2015.pdf | February 23, 2023 |
| 325XXXH01 - Rev A - Thermo Scientific PEEK - CO2 Back Up System Installation Instructions.pdf | November 22, 2021 |