HC Models TSX TLE RLE HLE 89000 Series ULT Freezers - WIFI Analyzer Instruction Sheet
This instruction sheet guides users on how to check the signal strength of a WiFi access point using a WiFi analyzer app on their phone. It includes steps to download the appropriate app from Google Play or the Apple Store and emphasizes that only 2.4GHz wireless is supported. Users are advised to compare the signal strength to the recommended levels provided in the chart. The document applies to new HC models: TSX, TLE, RLE, HLE, and 89000.
Description:
How to check signal strength with a WIFI analyzer app on phone
Environment:
- TSX
- TLE
- RLE
- HLE
- 89000
- Advanced Series
| 8930086A | HLE30086A | RLE30086A | TLE30086A | TSX40086A |
| 8930086D | HLE30086D | RLE30086D | TLE30086D | TSX40086D |
| 8930086V | HLE30086V | RLE30086V | TLE30086V | TSX40086V |
| 8940086A | HLE40086A | RLE40086A | TLE40086A | TSX40086G |
| 8940086D | HLE40086D | RLE40086D | TLE40086D | TSX50086A |
| 8940086V | HLE40086V | RLE40086V | TLE40086V | TSX50086D |
| 8950086A | HLE50086A | RLE50086A | TLE50086A | TSX50086V |
| 8950086D | HLE50086D | RLE50086D | TLE50086D | TSX50086G |
| 8950086V | HLE50086V | RLE50086V | TLE50086V | TSX60086A |
| 8960086A | HLE60086A | RLE60086A | TLE60086A | TSX60086D |
| 8960086D | HLE60086D | RLE60086D | TLE60086D | TSX60086V |
| 8960086V | HLE60086V | RLE60086V | TLE60086V |
TSX60086G |
| TSX70086D | TSX70086V | TSX70086G | TSX70086A |
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific TSX and HC Series ULT Freezers - WIFI Analyzer Instruction Sheet.pdf | November 22, 2021 |