PEEK and TSX - Restore to Factory Settings Instruction Sheet
Instruction Sheet PEEK & TSX Restore to factory settings 1. Click settings icon. 2. Select preference icon. 2 3. Press factory reset button. Once this is pressed unit will reboot as if it were brand new and have all default settings. If past data temperature or event logs are needed for records download on USB before completing. PEEK & TSK Instruction Sheet.
Description:
Instruction sheet to restore factory settings. Completing factory reset on PEEK and TSX Series Gen 1.
Environment:
Peek and TSX Series User Interface Assy:
| UxF30086A | HFU300TA | 88300A | TSU300A |
| UxF30086D | HFU300TD | 88300D | TSU300D |
| UxF30086V | HFU300TV | 88300V | TSU300V |
| UxF40086A | HFU400TA | 88400A | TSU400A |
| UxF40086D | HFU400TD | 88400D | TSU400D |
| UxF40086V | HFU400TV | 88400V | TSU400V |
| UxF50086A | HFU500TA | 88500A | TSU500A |
| UxF50086D | HFU500TD | 88500D | TSU500D |
| UxF50086V | HFU500TV | 88500V | TSU500V |
| UxF60086A | HFU600TA | 88600A | TSU600A |
| UxF60086D | HFU600TD | 88600D | TSU600D |
| UxF60086V | HFU600TV | 88600V | TSU600V |
| UxF70086D | HFU700TD | 88700D | TSU700D |
| UxF70086V | HFU700TV | 88700V | TSU700V |
| TSX400A | TSX400D | TSX400V | |
| TSX600A | TSX600D | TSX600V |
Attachment(s):
| File | Last Modified |
|---|---|
| PEEK and TSX - Restore to Factory Settings Instruction Sheet.pdf | November 22, 2021 |