TSX Core ULT Freezer Series - How to Adjust Display Brightness Instructions
This article explains how to adjust TSX Core Freezer display brightness via User Mode. It covers entering ‘usr’ (optional passcode; “Por” confirms activation), opening the Intensity (‘Int’) menu, and selecting 25%, 50%, 75%, or 100%. The chosen level saves automatically, persists after power cycles, and defaults to 75%. Lower settings help reduce glare in low-light environments.
Description:
Instruction Sheet on adjusting or changing Display Brightness on TSX Core Series.
Environment:
TSX Core Series, TSX40086CA , TSX50086CA . TSX60086CA , TSX70086CA , ISO40086GA , ISO40086GD , ISO40086GV , ISO50086GA , ISO50086GV , ISO60086GD , ISO60086GV , VWR40086GA , VWR40086GD , VWR40086GV , VWR50086GA , VWR50086GD , VWR50086GV , VWR60086GA , VWR60086GD , VWR60086GV , VWR70086GA, VWR70086GD , VWR70086GV
Attachment(s):
| File | Last Modified |
|---|---|
| TSX Core ULT Freezer Series - How to Adjust Dipslay Brightness Instructions.pdf | December 05, 2025 |