TSX Core ULT Freezer Series - How to Complete Factory Rest via Program Mode Instructions
This article provides instructions on performing a factory reset on a TSX Core ULT Freezer. It describes entering User Mode, navigating to the factory reset option, selecting and confirming the reset, and waiting for the system to reboot. Post-reset, the freezer's brightness resets to 75%, the passcode resets to 000, and all configuration values revert to default while logs remain intact. It emphasizes that these steps are essential for restoring default settings.
Description:
This article provide factory reset instructions for TSX Core Series via Program Mode.
Environment:
TSX Core Series, TSX40086CA , TSX50086CA . TSX60086CA , TSX70086CA , ISO40086GA , ISO40086GD , ISO40086GV , ISO50086GA , ISO50086GV , ISO60086GD , ISO60086GV , VWR40086GA , VWR40086GD , VWR40086GV , VWR50086GA , VWR50086GD , VWR50086GV , VWR60086GA , VWR60086GD , VWR60086GV , VWR70086GA, VWR70086GD , VWR70086GV
Attachment(s):
| File | Last Modified |
|---|---|
| TSX Core ULT Freezer Series - How to Complete Factory Rest via Program Mode Instructions.pdf | November 11, 2025 |