TSX Gen 2 HLE RLE TLE 89000 Series - ULT Freezers - Power on Reset Instruction Sheet
This instruction sheet provides steps for performing a power on reset (POR) on new HC models of TSX Gen 2 Series ULT Freezers to reboot the system and reconnect to WIFI and the cloud. It includes opening the bottom air filter door, disconnecting a battery wire, turning off and on the power switch, waiting 30 seconds, reconnecting the battery wire, and checking for WIFI and cloud icons on the display to confirm reconnection.
Description:
How to Perform a Power on Reset.
A power on reset (POR) is sometimes used to reboot the system. It can be used to reconnect the freezer to WIFI and the cloud.
Environment:
TSX Gen 2 Series, Advanced Series ULT Freezers
| 8930086A | HLE30086A | RLE30086A | TLE30086A | TSX40086A |
| 8930086D | HLE30086D | RLE30086D | TLE30086D | TSX40086D |
| 8930086V | HLE30086V | RLE30086V | TLE30086V | TSX40086G |
| 8940086A | HLE40086A | RLE40086A | TLE40086A | TSX40086V |
| 8940086D | HLE40086D | RLE40086D | TLE40086D | TSX50086A |
| 8940086V | HLE50086A | RLE40086V | TLE40086V | TSX50086D |
| 8950086A | HLE50086D | RLE50086A | TLE50086A | TSX50086G |
| 8950086D | HLE50086V | RLE50086D | TLE50086D | TSX50086V |
| 8950086V | HLE60086A | RLE50086V | TLE50086V | TSX60086A |
| 8960086A | HLE60086D | RLE60086A | TLE60086A | TSX60086D |
| 8960086D | HLE60086V | RLE60086D | TLE60086D | TSX60086G |
| 8960086V | RLE60086V | TLE60086V | TSX60086V | |
| TSX70086A | ||||
| TSX70086D | ||||
| TSX70086G | ||||
| TSX70086V |
Attachment(s):
| File | Last Modified |
|---|---|
| TSX HLE RLE TLE 89000 Series - ULT Freezers - Power on Reset Instruction Sheet.pdf | November 19, 2021 |