TSX Gen 2 HLE RLE TLE 89000 Series ULT Freezers - How to Shut Down and Restart Instruction Sheet
This article provides best practices for shutting down and restarting TSX ULT Freezers, particularly during facility closures due to the COVID-19 pandemic. It outlines steps to prevent battery depletion and water damage during shutdown, such as unplugging the freezer, leaving doors open for 48 hours, and drying the interior. For restarting, it includes plugging in the freezer, turning on the power, completing initial setup, and allowing 8-12 hours to reach -80°C. These guidelines ensure proper m
Description:
Best Practice for shutting down and restarting a TSX ULT Freezer
Environment:
TSX Gen 2 Series, RLE, 89000, HLE, and TLE Advanced series ULT Freezer proper power down.
| RLE30086A | 8930086A | HLE30086A | TLE30086A |
| RLE30086D | 8930086D | HLE30086D | TLE30086D |
| RLE30086V | 8930086V | HLE30086V | TLE30086V |
| RLE40086A | 8940086A | HLE40086A | TLE40086A |
| RLE40086D | 8940086D | HLE40086D | TLE40086D |
| RLE40086V | 8940086V | HLE40086V | TLE40086V |
| RLE50086A | 8950086A | HLE50086A | TLE50086A |
| RLE50086D | 8950086D | HLE50086D | TLE50086D |
| RLE50086V | 8950086V | HLE50086V | TLE50086V |
| RLE60086A | 8960086V | HLE60086A | TLE60086A |
| RLE60086D | 8960086D | HLE60086D | TLE60086D |
| RLE60086V | 8960086V | HLE60086V | TLE60086V |
| TSX40086A | TSX40086D | TSG40086V | TSX70086A |
| TSX50086A | TSX50086D | TSX50086V | TSX70086G |
| TSX60086A | TSX60086D | TSX60086V | TSX70086V |
| TSX70086D |
Attachment(s):
| File | Last Modified |
|---|---|
| TSX HLE RLE TLE 89000 Series ULT Freezers - How to Shut Down and Restart Instruction Sheet.pdf | November 22, 2021 |