TSX ISO Core ULT Freezer Series - How to Power Off Instructions
This article explains how to power off the TSX/ISO Core Freezer Series: enter User Mode (enter passcode if enabled), confirm “Por,” change shutdown from no to yes, start the 15-second countdown, then switch off the front power. It details the use of ✓ and +/– arrows, passcode entry, and the final power switch step for complete shutdown and device safety.
Description:
Instruction sheet on how to power off or shut down TSX and ISO Core ULT Freezer Series.
Environment:
TSX Core Series, TSX40086CA , TSX50086CA . TSX60086CA , TSX70086CA , ISO40086GA , ISO40086GD , ISO40086GV , ISO50086GA , ISO50086GV , ISO60086GD , ISO60086GV , VWR40086GA , VWR40086GD , VWR40086GV , VWR50086GA , VWR50086GD , VWR50086GV , VWR60086GA , VWR60086GD , VWR60086GV , VWR70086GA, VWR70086GD , VWR70086GV
Attachment(s):
| File | Last Modified |
|---|---|
| TSX ISO Core ULT Freezer Series - How to Power Off Instructions.pdf | December 05, 2025 |