Thermo Fisher Scientific
TSX Universal ULT Freezer Series - How Export Log Instructions
This guide explains how to export logs from the TSX Universal ULT Freezer Series. Insert a USB drive, select the log type (Event and Temperature, Event, or Temperature), choose the export format (PDF, CSV, or PUC), select a date range, and press "Export Log." Wait for the export to complete, then remove the USB drive and verify the files on a computer.
Description: 
The attached document provides step by step procedure on downloading export logs. 
Environment:
TSX Universal
  • TSX40086LA 
  • TSX50086LA
  • TSX60086LA
  • TSX70086LA
  • TSX40086FA
  • TSX50086FA
  • TSX60086FA
  • TSX70086FA 
  • Was this article helpful?