Thermo Scientific TSX TLE RLE HLE 89000 ULT Freezers - Factory Reset Instruction Sheet
This instruction sheet outlines the steps to perform a factory reset on models TSX, TLE, RLE, HLE, and 89000. First, access the settings, then navigate to Files and Info, and select Factory Reset. Acknowledge the reset to erase all customer and setup data from the device while retaining PUC and Log files saved in the UI.
Description:
How to do a factory reset and restore default settings on TSX and Advanced Series Freezers.
Environment:
TSX Gen 2 and Advanced Series TLE, RLE, HLE, and 89000 -86 freezers.
| RLE30086A | 8930086A | HLE30086A | TLE30086A |
| RLE30086D | 8930086D | HLE30086D | TLE30086D |
| RLE30086V | 8930086V | HLE30086V | TLE30086V |
| RLE40086A | 8940086A | HLE40086A | TLE40086A |
| RLE40086D | 8940086D | HLE40086D | TLE40086D |
| RLE40086V | 8940086V | HLE40086V | TLE40086V |
| RLE50086A | 8950086A | HLE50086A | TLE50086A |
| RLE50086D | 8950086D | HLE50086D | TLE50086D |
| RLE50086V | 8950086V | HLE50086V | TLE50086V |
| RLE60086A | 8960086V | HLE60086A | TLE60086A |
| RLE60086D | 8960086D | HLE60086D | TLE60086D |
| RLE60086V | 8960086V | HLE60086V | TLE60086V |
| TSX40086A | TSX40086D | TSG40086V | TSX70086A |
| TSX50086A | TSX50086D | TSX50086V | TSX70086G |
| TSX60086A | TSX60086D | TSX60086V | TSX70086V |
| TSX70086D |
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific TSX TLE RLE HLE 89000 ULT Freezers - Factory Reset Instruction Sheet.pdf | November 22, 2021 |