Thermo Scientific ULT Freezers 89000 HLE TLE RLE and TSX Series - Battery Replacement Instructions
This instruction sheet guides users on changing the battery in TSX Gen 2 Series ULT Freezers, including models TSX Gen 2, RLE, TLE, HLE, and 89000. It details steps such as opening the bottom door, locating and labeling the battery leads, disconnecting and removing the old battery, installing the new battery, and allowing 4-6 days for it to charge. Additionally, it includes instructions for resetting the battery clock by pressing specific buttons on the unit.
Description:
How to change or replace the battery on TSX Gen 2 and Advanced 89000, HLE, RLE, and TLE Series ULT Freezers.
Environment:
ULT Freezers, models:
| 8930086A | HLE30086A | RLE30086A | TLE30086A | TSX40086A |
| 8930086D | HLE30086D | RLE30086D | TLE30086D | TSX40086D |
| 8930086V | HLE30086V | RLE30086V | TLE30086V | TSX40086G |
| 8940086A | HLE40086A | RLE40086A | TLE40086A | TSX40086V |
| 8940086D | HLE40086D | RLE40086D | TLE40086D | TSX50086A |
| 8940086V | HLE40086V | RLE40086V | TLE40086V | TSX50086D |
| 8950086A | HLE50086A | RLE50086A | TLE50086A | TSX50086G |
| 8950086D | HLE50086D | RLE50086D | TLE50086D | TSX50086V |
| 8950086V | HLE50086V | RLE50086V | TLE50086V | TSX60086A |
| 8960086A | HLE60086A | RLE60086A | TLE60086A | TSX60086D |
| 8960086D | HLE60086D | RLE60086D | TLE60086D | TSX60086G |
| 8960086V | HLE60086V | RLE60086V | TLE60086V | TSX60086V |
| TSX70086A | ||||
| TSX70086D | ||||
| TSX70086V |
Attachment(s):
| File | Last Modified |
|---|---|
| TSX Gen 2 and Advanced Series ULT Freezers - How to Change Battery Instruction Sheet.pdf | September 18, 2025 |
| Thermo Scientific ULT Freezers 89000 HLE TLE RLE and TSX Series - Battery Replacement Instrucitons.pdf | October 04, 2023 |