Thermo Scientific Cryo Preservation System - 7400 Cryo Plus 90L Storage System - Technical Data Sheet
This specification sheet describes the Thermo Scientific CryoPlus 7400, a 90L liquid nitrogen cryogenic storage system for long-term biological sample storage. It covers intended use, CFDA/cULus/CE listings, 100/120V NEMA 5-15P power, vacuum-insulated stainless chamber, microprocessor controller, auto-fill, level LEDs, remote alarm, up to 6,318 vials, dimensions/weights, -135°C to -196°C range, 3 L/day evaporation, and warranty.
Description:
Cold Storage specification sheet. Also known as a technical data sheet.
Environment:
Thermo Scientific Cryo Preservation System, Model 7400, Cryo Plus 90L Storage system
Attachment(s):
| File | Last Modified |
|---|---|
| Rev B - Thermo Scientific Cryo Preservation System, Model 7400, Cryo Plus 90L Storage System - Technical Data Sheet.pdf | December 10, 2021 |