Thermo Scientific Cryo Preservation System - 7402 Cryo Plus 200L Storage System - Technical Data Sheet
This specification sheet describes the Thermo Scientific CryoPlus 200L (Model 7402), a liquid nitrogen cryogenic storage system for long-term biological sample preservation. Key specs: 200L LN2, vapor phase −135°C to −196°C, up to 13,500 2 mL vials, vacuum-insulated stainless chamber, microprocessor control, auto-fill with hi/low setpoints, 24-LED level monitor, remote alarm, 100/120V 50/60Hz, 0.8A, NEMA 5-15P, CFDA/cULus/CE, 34×28.5×41 in, ~5 L/day SER, keyed locks.
Description:
7402 specification sheet. Alson known as a technical data sheet.
Environment:
Thermo Scientific Cryo Preservation System, Model 7402, Cryo Plus 200L Storage System
Attachment(s):
| File | Last Modified |
|---|---|
| Rev B - Thermo Scientific Cryo Preservation System, Model 7402, Cryo Plus 200L Storage System - Technical Data Sheet.pdf | December 10, 2021 |