Thermo Scientific Cryo Preservation System - 7403 Cryo Plus 200L Storage System - Technical Data Sheet
Thermo Scientific CryoPlus 200L (7403) cryo preservation system specification sheet for long-term biological sample storage in vapor-phase liquid nitrogen. Details capacity (200L, 13,500 vials), temperature range (-135°C to -196°C), auto-fill, LED level monitoring, microprocessor control, security locks, electrical requirements, dimensions, weights, and compliance/warranty info.
Description:
Model 7403 specification sheet. Alson known as a technical data sheet.
Environment:
Thermo Scientific Cryo Preservation System, Model 7403, Cryo Plus 200L Storage System
Attachment(s):
| File | Last Modified |
|---|---|
| Rev B - Thermo Scientific Cryo Preservation System, Model 7403, Cryo Plus 200L Storage System - Technical Data Sheet.pdf | December 10, 2021 |