Thermo Scientific Cryo Preservation System - Model 7401 Cryo Plus 90L Storage System - Technical Data Sheet
This specification sheet details the Thermo Scientific CryoPlus 7401, a 90L liquid nitrogen cryogenic storage system for long-term biological sample preservation. It outlines intended use, CFDA/cULus/CE listings, 200/230V CEE 7/7 power, microprocessor controller, auto-fill with level LEDs, -135°C to -196°C vapor phase, 3 L/day evaporation, 6,318-vial capacity, dimensions/weights, vacuum-insulated stainless chamber, and remote alarm.
Description:
Cold Storage specification sheet. Also known as a technical data sheet.
Environment:
Thermo Scientific Cryo Preservation System, Model 7401, Cryo Plus 90L Storage System
Attachment(s):
| File | Last Modified |
|---|---|
| Rev B - Thermo Scientific Cryo Preservation System, Model 7401, Cryo Plus 90L Storage System - Technical Data Sheet.pdf | December 10, 2021 |