Thermo Scientific - TSX1 HP Flex Laboratory Freezer - TSX1-2530FA - Technical Data Sheet
Technical specifications for the Thermo Scientific TSX1 HP Flex Auto Defrost Freezer, model TSX1-2530FA, featuring a solid door and 25 cu. ft. (707.9 L) capacity for laboratory storage at -35C to -15C. Operates on 115V, 60Hz, single-phase power at 6.3A. Features -30C setpoint, touchscreen control, automatic defrost, R290 refrigerant, adjustable temperature alarms, power failure alarm, 24-hour/7-day temperature history, RS485 connectivity, USB data export, and remote alarm contacts.
Description:
Attached is the technical data sheet also known as a specification sheet for model TSX1-2530FA.
Environment:
TSX1 HP Flex, TSX1 Flex Series, TSX1 Flex High Performance Freezer, TSX1-2530FA, specs, tds
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific - TSX1 HP Flex Laboratory Freezer - TSX1-2530FA - Technical Data Sheet.pdf | October 23, 2026 |